Skip to Content
Merck
All Photos(1)

Key Documents

SML1807

Sigma-Aldrich

SB-258719

≥98% (HPLC)

Synonym(s):

N,3-Dimethyl-N-[(1R)-1-methyl-3-(4-methyl-1-piperidinyl)propyl]benzenesulfonamide, SB 258719

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$694.00
25 MG
R$2,791.00

R$694.00


Estimated to ship onMay 21, 2025



Select a Size

Change View
5 MG
R$694.00
25 MG
R$2,791.00

About This Item

Empirical Formula (Hill Notation):
C18H30N2O2S
CAS Number:
Molecular Weight:
338.51
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

R$694.00


Estimated to ship onMay 21, 2025


Quality Level

Assay

≥98% (HPLC)

form

oil

color

off-white to brown

storage temp.

2-8°C

SMILES string

CC1=CC=CC(S(N(C)[C@H](C)CCN2CCC(C)CC2)(=O)=O)=C1

InChI

1S/C18H30N2O2S/c1-15-8-11-20(12-9-15)13-10-17(3)19(4)23(21,22)18-7-5-6-16(2)14-18/h5-7,14-15,17H,8-13H2,1-4H3

InChI key

AGVNHDNTFYHZNL-UHFFFAOYSA-N

Biochem/physiol Actions

SB-258719 is a selective and potent 5-HT7 antagonist. SB-258719 reverses the hypothermic effect of 5-CT in mice.
Selective and potent 5-HT7 antagonist

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xin Zhao et al.
Scientific reports, 5, 9043-9043 (2015-03-13)
Fisetin, a natural flavonoid, has been shown in our previous studies to exert antidepressant-like effect. As antidepressant drugs are clinically used to treat chronic neuropathic pain, this work aimed to investigate the potential antinociceptive efficacies of fisetin against neuropathic pain
D R Thomas et al.
British journal of pharmacology, 124(6), 1300-1306 (1998-08-28)
1. The functional profile of the long form of the human cloned 5-HT7 receptor (designated h5-HT7(a)) was investigated using a number of 5-HT receptor agonists and antagonists and compared with its binding profile. Receptor function was measured using adenylyl cyclase
Sarwat Fatima et al.
Molecular oncology, 10(2), 195-212 (2015-10-18)
5-Hydroxytryptamine (5-HT), a neurotransmitter and vasoactive factor, has been reported to promote proliferation of serum-deprived hepatocellular carcinoma (HCC) cells but the detailed intracellular mechanism is unknown. As Wnt/β-catenin signalling is highly dysregulated in a majority of HCC, this study explored

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service