Skip to Content
Merck
All Photos(1)

Key Documents

SML1799

Sigma-Aldrich

UCPH-102

≥98% (HPLC)

Synonym(s):

2-Amino-4-methyl-7-(naphthalen-1-yl)-5-oxo-5,6,7,8-tetrahydro-4H-chromene-3-carbonitrile, 2-Amino-5,6,7,8-tetrahydro-4-methyl-7-(1-naphthalenyl)-5-oxo-4H-1-benzopyran-3-carbonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$1,022.00

R$1,022.00


Estimated to ship onMay 28, 2025



Select a Size

Change View
5 MG
R$1,022.00

About This Item

Empirical Formula (Hill Notation):
C21H18N2O2
CAS Number:
Molecular Weight:
330.38
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

R$1,022.00


Estimated to ship onMay 28, 2025


Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1)C2=C(OC(N)=C(C#N)C2C)CC1C3=C(C=CC=C4)C4=CC=C3

InChI key

XZQMHUGTNOOYFX-UHFFFAOYSA-N

Biochem/physiol Actions

UCPH-102 is a selective inhibitor of the Excitatory Amino Acid Transporter subtype-1 (EAAT1, GLAST) with an IC50 value of 420 nM for EAAT1 and >300 μM for EAAT2-5. UCPH-102 has been shown to cross the blood-brain barrier, so should become a valuable tool for studying the role of EAAT1 in brain.
UCPH-102 is a selective inhibitor of the Excitatory Amino Acid Transporter subtype-1 (EAAT1, GLAST).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bjarke Abrahamsen et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(3), 1068-1087 (2013-01-18)
In the present study, the mechanism of action and molecular basis for the activity of the first class of selective inhibitors of the human excitatory amino acid transporter subtype 1 (EAAT1) and its rodent ortholog GLAST are elucidated. The previously
Tri H V Huynh et al.
Bioorganic & medicinal chemistry, 20(23), 6831-6839 (2012-10-18)
The excitatory amino acid transporters (EAATs) play a pivotal role in regulating the synaptic concentration of glutamate in the mammalian central nervous system. To date, five different subtypes have been identified, named EAAT15 in humans (and GLAST, GLT-1, EAAC1, EAAT4
Mette N Erichsen et al.
Neurochemical research, 39(10), 1964-1979 (2014-04-01)
Uptake of the major excitatory neurotransmitter in the CNS, (S)-glutamate, is mediated by a family of excitatory amino acid transporters (EAAT). Previously we have explored the structure-activity relationship (SAR) of a series of EAAT1 selective inhibitors, leading to the development

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service