Skip to Content
Merck
All Photos(1)

Key Documents

SML1726

Sigma-Aldrich

MSAB

≥95% (HPLC)

Synonym(s):

3-[[(4-Methylphenyl)sulfonyl]amino]-benzoic acid methyl ester, Methyl 3-{[(4-methylphenyl)sulfonyl]amino}benzoate, WI-21

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$694.00
25 MG
R$2,791.00

R$694.00


Check Cart for Availability


Select a Size

Change View
5 MG
R$694.00
25 MG
R$2,791.00

About This Item

Empirical Formula (Hill Notation):
C15H15NO4S
CAS Number:
Molecular Weight:
305.35
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

R$694.00


Check Cart for Availability

Quality Level

Assay

≥95% (HPLC)

form

powder

color

off-white to purple

solubility

DMSO: 25 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC1=CC=C(S(NC2=CC(C(OC)=O)=CC=C2)(=O)=O)C=C1

InChI key

CVKBYFCJQSPBOI-UHFFFAOYSA-N

Biochem/physiol Actions

A cell-permeable compound that targets β-catenin via direct affinity interaction within the C-terminal two thirds (a.a. 301-670) of the Armadillo repeat region (a.a. 136–686), inducing β-catenin ubiquitination and proteasomal degradation (Eff. conc. 1.25-5 μM in DLD-1, SW480 and LS174T cultures). MSAB selectively inhibits against Wnt sginaling-dependent proliferation of cancer cells (IC50 <6 μM), while exhibiting little efficacy in Wnt-independent cultures (<10% inhibition at 10 μM). Likewise, daily intraperitoneal injection (10-20 mg/kg/day) is efficacious in suppressing the expansion of established tumors from HCT116, HT115, and H23 xenografts in mice, while exhibiting little efficacy against the Wnt-independent H460 tumor growth.
A cellular β-catenin degradation inducer with anti-proliferation efficacy against Wnt-dependent cancer growth both in vitro and in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Young-Eun Cho et al.
International journal of molecular sciences, 23(16) (2022-08-27)
Zinc finger protein with KRAB and SCAN domains 3 (ZKSCAN3) acts as an oncogenic transcription factor in human malignant tumors, including colon and prostate cancer. However, most of the ZKSCAN3-induced carcinogenic mechanisms remain unknown. In this study, we identified ZKSCAN3
Peder R Braadland et al.
Molecular cancer research : MCR, 17(11), 2154-2168 (2019-08-10)
The incidence of treatment-related neuroendocrine prostate cancer (t-NEPC) is rising as more potent drugs targeting the androgen signaling axis are clinically implemented. Neuroendocrine transdifferentiation (NEtD), an putative initial step in t-NEPC development, is induced by androgen-deprivation therapy (ADT) or anti-androgens
Cody J Aros et al.
Cell reports, 30(7), 2055-2064 (2020-02-23)
Mechanisms underpinning airway epithelial homeostatic maintenance and ways to prevent its dysregulation remain elusive. Herein, we identify that β-catenin phosphorylated at Y489 (p-β-cateninY489) emerges during human squamous lung cancer progression. This led us to develop a model of airway basal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service