Skip to Content
Merck
All Photos(1)

Key Documents

SML1722

Sigma-Aldrich

LUF5834

≥98% (HPLC)

Synonym(s):

2-Amino-4-(4-hydroxyphenyl)-6-(1H-imidazol-2-ylmethylsulfanyl)pyridine-3,5-dicarbonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$638.00
25 MG
R$2,568.00

R$638.00


Check Cart for Availability


Select a Size

Change View
5 MG
R$638.00
25 MG
R$2,568.00

About This Item

Empirical Formula (Hill Notation):
C17H12N6OS
CAS Number:
Molecular Weight:
348.38
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

R$638.00


Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

NC1=C(C#N)C(C2=CC=C(O)C=C2)=C(C#N)C(SCC3N=CC=N3)=N1

InChI

1S/C17H12N6OS/c18-7-12-15(10-1-3-11(24)4-2-10)13(8-19)17(23-16(12)20)25-9-14-21-5-6-22-14/h1-6,24H,9H2,(H2,20,23)(H,21,22)

InChI key

OFHKDLYFKPBXER-UHFFFAOYSA-N

Biochem/physiol Actions

LUF5834 is a potent A2A and A2B adenosine receptor partial agonist with an EC50 value of 12 nM and 45-fold selectivity over the adenosine A3 receptor.
Potent A2A and A2B adenosine receptor partial agonist

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service