Skip to Content
Merck
All Photos(1)

Key Documents

SML1691

Sigma-Aldrich

ZM323881 hydrochloride

≥98% (HPLC)

Synonym(s):

5-((7-Benzyloxyquinazolin-4-yl)amino)-4-fluoro-2-methylphenol hydrochloride, ZM-323881 hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$813.00
25 MG
R$2,878.00

R$813.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
R$813.00
25 MG
R$2,878.00

About This Item

Empirical Formula (Hill Notation):
C22H18FN3O2 · HCl
CAS Number:
Molecular Weight:
411.86
UNSPSC Code:
12352200
NACRES:
NA.77

R$813.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

Fc1c(cc(c(c1)C)O)Nc2ncnc3c2ccc(c3)OCc4ccccc4.Cl

InChI

1S/C22H18FN3O2.ClH/c1-14-9-18(23)20(11-21(14)27)26-22-17-8-7-16(10-19(17)24-13-25-22)28-12-15-5-3-2-4-6-15;/h2-11,13,27H,12H2,1H3,(H,24,25,26);1H

InChI key

AVRHWGLIYGJSOD-UHFFFAOYSA-N

Biochem/physiol Actions

ZM323881 is a selective inhibitor of VEGFR-2 with an IC50 value of 2 nM. ZM323881 is selective for VEGFR-2 over VEGFR-1 and other receptor tyrosine kinases including PDGFRβ, FGFR1, EGFR and erbB2. ZM323881 inhibited VEGF-A-induced endothelial cell proliferation with an IC50 value of 8 nM.
ZM323881 is a selective inhibitor of VEGFR-2.
ZM323881 is also known as (5‐{[7‐(benzyloxy) quinazolin‐4‐yl]amino}‐4‐fluoro‐2‐methylphenol).[1] It is capable of blocking the efflux function and altering breast cancer resistant protein (BCRP)-related multidrug resistance (MDR).[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C E Whittles et al.
Microcirculation (New York, N.Y. : 1994), 9(6), 513-522 (2002-12-17)
Vascular endothelial growth factor (VEGF) increases vascular permeability and angiogenesis in many pathological conditions including cancer, arthritis, and diabetes. VEGF activates VEGF-Receptor 1(VEGF-R1) and VEGF-Receptor 2 (VEGF-R2), which autophosphorylate to initiate a signaling cascade resulting in angiogenesis and increased microvascular
Selective reversal of BCRP-mediated MDR by VEGFR-2 inhibitor ZM323881
Zhang Yun-Kai, et al.
Biochemical Pharmacology, 132, 29-37 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service