Skip to Content
Merck
All Photos(1)

Key Documents

SML1689

Sigma-Aldrich

Eupatilin

from (Isolated from Artemisia sp.), ≥98% (HPLC), powder, lipophilic flavonoid

Synonym(s):

2-(3,4-Dimethoxyphenyl)-5,7-dihydroxy-6-methoxy-4H-1-benzopyran-4-one, 5,7-Dihydroxy-3′,4′,6-trimethoxyflavone, NSC 122413

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$766.00
25 MG
R$3,075.00

R$766.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
R$766.00
25 MG
R$3,075.00

About This Item

Empirical Formula (Hill Notation):
C18H16O7
CAS Number:
Molecular Weight:
344.32
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

R$766.00


Check Cart for Availability

Request a Bulk Order

Product Name

Eupatilin, ≥98% (HPLC)

biological source

(Isolated from Artemisia sp.)

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated
protect from light

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C1C=C(C2=CC=C(OC)C(OC)=C2)OC3=CC(O)=C(OC)C(O)=C31

InChI

1S/C18H16O7/c1-22-12-5-4-9(6-14(12)23-2)13-7-10(19)16-15(25-13)8-11(20)18(24-3)17(16)21/h4-8,20-21H,1-3H3

InChI key

DRRWBCNQOKKKOL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Eupatilin has been used to investigate its inhibitory effects on intracranial hemorrhage (ICH)-induced inflammation and its anti-inflammatory mechanisms in mouse BV2 cells, in vitro.[1] It has also been used as a standard for its quantitative estimation in Artemisia extracts by microcolumn high performance liquid chromatography with diode array (HPLC-DAD) assay.[2]

Biochem/physiol Actions

Eupatilin is a flavone with anti-oxidative, anti-inflammatory, and anti-cancer properties.
Eupatilin is an important constituent in the leaves of Artemisia argyi and also an active component of DA-9601.[3] It exhibits anti-apoptotic effects. Eupatilin provides protection against tumor necrosis factor α (TNF-α)-mediated inflammation in human umbilical vein endothelial cells.[1] It is used in the treatment of gastritis and peptic ulcers.[3] Eupatilin safeguards gastric epithelial cells from oxidative damage and smooth muscle cells from indomethacin-induced cell damage. Eupatilin lowers bile acid-stimulated hepatocyte apoptosis.[4]
Flavone with anti-oxidative, anti-inflammatory, and anti-cancer properties

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eupatilin attenuates the inflammatory response induced by intracerebral hemorrhage through the TLR4/MyD88 pathway
Fei X, et al.
International Immunopharmacology, 76, 105837-105837 (2019)
Eupatilin inhibits lipopolysaccharide-induced expression of inflammatory mediators in macrophages
Choi EJ, et al.
Life Sciences, 88(25-26), 1121-1126 (2011)
Yawei Wang et al.
Molecular medicine reports, 13(2), 1141-1146 (2015-12-18)
Eupatilin, one of the major flavonoids in Artemisia asiatica Nakai (Asteraceae), has been reported to possess antitumor properties. However, thus far there have been no reports regarding the effects of eupatilin on glioma. Therefore, in the current study the effects
Bioactive phenolics of the genus Artemisia (Asteraceae): HPLC-DAD-ESI-TQ-MS/MS profile of the Siberian species and their inhibitory potential against alpha-amylase and alpha-glucosidase
Olennikov DN, et al.
Frontiers in Pharmacology, 9, 756-756 (2018)
Antonia Wiegering et al.
Molecular biology of the cell, 32(8), 675-689 (2021-02-25)
A range of severe human diseases called ciliopathies is caused by the dysfunction of primary cilia. Primary cilia are cytoplasmic protrusions consisting of the basal body (BB), the axoneme, and the transition zone (TZ). The BB is a modified mother

Questions

  1. Peut on congeler et décongeler plusieurs fois l'eupatilin sans affecter son activité ?

    1 answer
    1. This product has not been analyzed for stability with repetitive freeze thaw cycles. Kindly generate aliquots to avoid product failure caused by repeated freezing and thawing.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service