Skip to Content
Merck
All Photos(1)

Key Documents

SML1681

Sigma-Aldrich

Endosidin2

≥98% (HPLC)

Synonym(s):

(E)-3-Fluoro-N′-(4-hydroxy-3-iodo-5-methoxybenzylidene)benzohydrazide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$789.00
25 MG
R$3,181.00

R$789.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
R$789.00
25 MG
R$3,181.00

About This Item

Empirical Formula (Hill Notation):
C15H12FIN2O3
CAS Number:
Molecular Weight:
414.17
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

R$789.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC1=CC(C(N/N=C/C2=CC(OC)=C(O)C(I)=C2)=O)=CC=C1

InChI

1S/C15H12FIN2O3/c1-22-13-6-9(5-12(17)14(13)20)8-18-19-15(21)10-3-2-4-11(16)7-10/h2-8,20H,1H3,(H,19,21)/b18-8+

InChI key

UIADDCURKKFTFW-QGMBQPNBSA-N

Biochem/physiol Actions

Endosidin2 is a cell-permeable benzylidene-benzohydrazide based compound that binds to the exocyst component of 70 kDa (EXO70) subunit of exocyst complex (Kd = 253 μM for interaction with EXO70A1) and reduces exocytosis and endosomal recycling in plant and mammalian cells. Displays good aqueous stability. Endosidin2 has shown to disrupt the trafficking of proteins between the plasma membrane and endosome, which leads to enhanced trafficking to the vacuole for degradation. It is reported to inhibit the recycling of endocytosed transferrin to plasma membrane in HeLa cells, and can target multiple isoforms of EXO70 in mammals, resulting in misregulation of exocytosis.  
Endosidin2 is a cell-permeable benzylidene-benzohydrazide based compound that binds to the exocyst component of 70 kDa (EXO70) subunit of exocyst complex and reduces exocytosis and endosomal recycling in plant and mammalian cells.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service