Skip to Content
Merck
All Photos(1)

Key Documents

SML1615

Sigma-Aldrich

SEN177

≥98% (HPLC)

Synonym(s):

6′′-Fluoro-4-(4-methyl-4H-[1,2,4]triazol-3-yl)-3,4,5,6-tetrahydro-2H-[1,2′;3′,3′′]terpyridine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$989.00
25 MG
R$3,979.00

R$989.00


Estimated to ship onApril 30, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
R$989.00
25 MG
R$3,979.00

About This Item

Empirical Formula (Hill Notation):
C18H19FN6
CAS Number:
Molecular Weight:
338.38
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

R$989.00


Estimated to ship onApril 30, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

FC(C=C1)=NC=C1C2=CC=CN=C2N(CC3)CCC3C4=NN=CN4C

InChI

1S/C18H19FN6/c1-24-12-22-23-17(24)13-6-9-25(10-7-13)18-15(3-2-8-20-18)14-4-5-16(19)21-11-14/h2-5,8,11-13H,6-7,9-10H2,1H3

InChI key

AJIAMIPUWJQSPR-UHFFFAOYSA-N

Biochem/physiol Actions

SEN177 is a Glutaminyl cyclase (QPCT) inhibitor.
SEN177 is a Glutaminyl cyclase (QPCT) inhibitor. Glutaminyl cyclase modifies N-terminal glutamine or glutamate residues to N-terminal 5-oxoproline or pyroglutamate (named QPCT). QPCT inhibits mutant huntingtin from forming aggregates, but also reduces aggregation of other aggregate-prone proteins containing polyQ or polyA repeats. SEN177 caused dose-dependent decreases in HTT aggregation by a mechanism that requires QPCT inhibition. SEN177 had in vitro activity against polyglutamine toxicity and was able to reduce aggregates in mammalian cells, primary neurons and in a Drosophila model.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service