Skip to Content
Merck
All Photos(1)

Key Documents

SML1398

Sigma-Aldrich

TM5275 sodium salt

≥98% (HPLC)

Synonym(s):

2-[2-[2-(4-Benzhydrylpiperazin-1-yl)-2-oxoethoxy]acetamido]-5-chlorobenzoic acid sodium salt, 5-Chloro--[[2-[2-[4-(diphenylmethyl)-1-piperazinyl]-2-oxoethoxy]acetyl]amino]-benzoic acid sodium salt, Sodium 5-chloro-2-[({2-[4-(diphenylmethyl) piperazin-1-yl]-2-oxoethoxy}acetyl)amino]benzoate, TM 5275, TM-5275

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$1,584.00
25 MG
R$5,618.00

R$1,584.00


Check Cart for Availability


Select a Size

Change View
5 MG
R$1,584.00
25 MG
R$5,618.00

About This Item

Empirical Formula (Hill Notation):
C28H27ClN3O5 · Na
CAS Number:
Molecular Weight:
543.97
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

R$1,584.00


Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(COCC(NC1=C(C([O-])=O)C=C(Cl)C=C1)=O)N2CCN(C(C3=CC=CC=C3)C4=CC=CC=C4)CC2.[Na+]

InChI

1S/C28H28ClN3O5.Na/c29-22-11-12-24(23(17-22)28(35)36)30-25(33)18-37-19-26(34)31-13-15-32(16-14-31)27(20-7-3-1-4-8-20)21-9-5-2-6-10-21;/h1-12,17,27H,13-16,18-19H2,(H,30,33)(H,35,36);/q;+1/p-1

InChI key

JSHSGBIWNPQCQZ-UHFFFAOYSA-M

Biochem/physiol Actions

TMS5275 is an orally available, potent and selective inhibitor of PAI-1 that delivers antithrombotic benefits devoid of bleeding effect in nonhuman primates.
TMS5275 is an orally available, potent and selective inhibitor of PAI-1.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service