Skip to Content
Merck
All Photos(1)

Key Documents

SML1389

Sigma-Aldrich

Farrerol

≥98% (HPLC)

Synonym(s):

(-)-Farrerol, (S)-2,3-Dihydro-5,7-dihydroxy-2-(4-hydroxyphenyl)-6,8-dimethyl-4-benzopyrone, 4′,5,7-Trihydroxy 6,8-dimethylflavanone, CID 442396

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16O5
CAS Number:
Molecular Weight:
300.31
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC1=C(O)C(C)=C(O)C2=C1O[C@H](C3=CC=C(O)C=C3)CC2=O

InChI

1S/C17H16O5/c1-8-15(20)9(2)17-14(16(8)21)12(19)7-13(22-17)10-3-5-11(18)6-4-10/h3-6,13,18,20-21H,7H2,1-2H3/t13-/m0/s1

InChI key

DYHOLQACRGJEHX-ZDUSSCGKSA-N

Biochem/physiol Actions

Antibechic, anti-bacterial, anti-inflammatory
Farrerol is major bioactive component from Rhododendron dauricum L. (Man-Shan-Hong). Farrerol exhibits numerous activities including antibechic, anti-bacterial, anti-inflammatory and inhibition of vascular smooth muscle cells (VSMC) proliferation. Recent study shows that Farrerol regulates occludin expression by preventing H2O2-induced activation ERK 1/2 in human endothelium-derived EA.hy926 cells.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weina Zhang et al.
eLife, 9 (2020-07-10)
Directly modulating the choice between homologous recombination (HR) and non-homologous end joining (NHEJ) - two independent pathways for repairing DNA double-strand breaks (DSBs) - has the potential to improve the efficiency of gene targeting by CRISPR/Cas9. Here, we have developed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service