Skip to Content
Merck
All Photos(1)

Documents

SML1384

Sigma-Aldrich

BEC hydrochloride

≥98% (HPLC)

Synonym(s):

S-(2-Boronoethyl)-L-cysteine, S-(2-Boronoethyl)-L-cysteine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12BNO4S · xHCl
CAS Number:
Molecular Weight:
193.03 (free base basis)
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

OB(O)CCSC[C@H](N)C(O)=O.C

InChI

1S/C5H12BNO4S.CH4/c7-4(5(8)9)3-12-2-1-6(10)11;/h4,10-11H,1-3,7H2,(H,8,9);1H4/t4-;/m0./s1

InChI key

IMYJQAMXIIYUIH-WCCKRBBISA-N

Biochem/physiol Actions

BEC (S-(2-boronoethyl)-l-cysteine) is a potent and specific arginase inhibitor that restores flow-induced responses in arterioles from diabetic rats.
BEC is a boronic analog of L-arginine, and L-cysteine. It competitively inhibits both arginase I and II. BEC does not directly affect nitric oxide synthase.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arginase inhibition restores arteriolar endothelial function in Dahl rats with salt-induced hypertension
Johnson FK, et al.
American Journal of Physiology. Lung Cellular and Molecular Physiology, 288(4), R1057-R1062 (2005)
Amino Acids in Human Nutrition and Health, 62-62 (2012)
Alessandra Romano et al.
Expert review of molecular diagnostics, 18(7), 675-683 (2018-05-01)
Despite improvement in overall response due to the introduction of the first-in-class proteasome inhibitor bortezomib (btz), multiple myeloma (MM) is still an incurable disease due to the immune-suppressive bone marrow (BM) environment. Thus, the authors aimed to identify the role
Barbara Érsek et al.
Cellular and molecular life sciences : CMLS, 78(2), 661-673 (2020-04-25)
This study shows that melanoma-associated fibroblasts (MAFs) suppress cytotoxic T lymphocyte (CTL) activity and reveals a pivotal role played by arginase in this phenomenon. MAFs and normal dermal fibroblasts (DFs) were isolated from surgically resected melanomas and identified as Melan-A-/gp100-/FAP+ cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service