Skip to Content
Merck
All Photos(1)

Key Documents

SML1383

Sigma-Aldrich

OG-L002 hydrochloride

≥98% (HPLC)

Synonym(s):

4′-((1R,2S)-2-Aminocyclopropyl)-[1,1′-biphenyl]-3-ol hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$1,120.00
25 MG
R$4,497.00

R$1,120.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
R$1,120.00
25 MG
R$4,497.00

About This Item

Empirical Formula (Hill Notation):
C15H15NO · HCl
CAS Number:
Molecular Weight:
261.75
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

R$1,120.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

, light yellow to dark orange

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

N[C@@H]1[C@@H](C(C=C2)=CC=C2C3=CC=CC(O)=C3)C1.Cl

InChI

1S/C15H15NO.ClH/c16-15-9-14(15)11-6-4-10(5-7-11)12-2-1-3-13(17)8-12;/h1-8,14-15,17H,9,16H2;1H/t14-,15+;/m1./s1

InChI key

LPVCAMIPTMRRLZ-LIOBNPLQSA-N

Biochem/physiol Actions

OG-L002 is a recently developed inhibitor of LSD1.
OG-L002 is a recently developed inhibitor with IC50 of 20 nM at LSD1 and much lower activity at MAO-A and MAO-B (1.38 and 0.72 μM, respectively). OG-L002 exhibits potent anti-viral activity in vitro and in mouse models of HSV infection. Lysine specific demethylase 1 (LSD1) is a histone demethylase that removes methyl groups from lysine 4 or 9 of H3 histone tails. Inhibition of LSD1 leds to suppression of herpes simplex and herpes zoster viral infections and viral reactivation from latency. MAO inhibitors (pargyline, tranylcypromine) are known to inhibit LSD1, but with low potency and selectivity. OG-L002 is a more selective and potent tool for LSD1 inhibition.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service