Skip to Content
Merck
All Photos(1)

Key Documents

SML1375

Sigma-Aldrich

Galeterone

≥98% (HPLC)

Synonym(s):

3β-Hydroxy-17-(1H-benzimidazole-1-yl)androsta-5,16-diene, TOK-001, VN-124-1, VN/124-1

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$715.00
25 MG
R$2,880.00

R$715.00


Estimated to ship onApril 14, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
R$715.00
25 MG
R$2,880.00

About This Item

Empirical Formula (Hill Notation):
C26H32N2O
CAS Number:
Molecular Weight:
388.55
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

R$715.00


Estimated to ship onApril 14, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

C[C@]12C(C[C@@H](O)CC2)=CC[C@]3([H])[C@]1([H])CC[C@@]4(C)[C@@]3([H])CC=C4N5C=NC6=C5C=CC=C6

InChI

1S/C26H32N2O/c1-25-13-11-18(29)15-17(25)7-8-19-20-9-10-24(26(20,2)14-12-21(19)25)28-16-27-22-5-3-4-6-23(22)28/h3-7,10,16,18-21,29H,8-9,11-15H2,1-2H3/t18-,19-,20-,21-,25-,26-/m0/s1

InChI key

PAFKTGFSEFKSQG-PAASFTFBSA-N

Gene Information

Biochem/physiol Actions

Galeterone is an inhibitor of CYP17A1 (CYP450c17), an enzyme that catalyzes two key serial reactions (17-α hydroxylase and 17,20 lyase) in androgen and estrogen biosynthesis resulting in the formation of DHEA and androstenedione, which may ultimately be metabolized into testosterone. CYP17 is the key enzyme for androgen biosynthesis in both the testes and adrenals, so its inhibition should stop the production of androgens in both places. Galeterone, moreover, is a potent blocker of the androgen receptor and causes down-regulation of androgen receptor (AR) protein expression in vitro and in vivo.
Galeterone is an inhibitor of CYP17A1.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service