Skip to Content
Merck
All Photos(1)

Key Documents

SML1339

Sigma-Aldrich

AR420626

≥98% (HPLC)

Synonym(s):

AR 420626, N-(2,5-Dichlorophenyl)-4-(furan-2-yl)-2-methyl-5-oxo-1,4,5,6,7,8-hexahydro-quinoline-3-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$899.00
25 MG
R$3,628.00

R$899.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
R$899.00
25 MG
R$3,628.00

About This Item

Empirical Formula (Hill Notation):
C21H18Cl2N2O3
CAS Number:
Molecular Weight:
417.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

R$899.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

ClC1=CC(NC(C2=C(C)NC(CCCC3=O)=C3C2C4=CC=CO4)=O)=C(Cl)C=C1

InChI

1S/C21H18Cl2N2O3/c1-11-18(21(27)25-15-10-12(22)7-8-13(15)23)20(17-6-3-9-28-17)19-14(24-11)4-2-5-16(19)26/h3,6-10,20,24H,2,4-5H2,1H3,(H,25,27)

InChI key

GGTYQECCGLBHGS-UHFFFAOYSA-N

Biochem/physiol Actions

AR420626 is a selective agonist of Free Fatty Acid 3 receptor FFA3 (previously called GPR41).
AR420626 is a selective agonist of Free Fatty Acid 3 receptor FFA3 (previously called GPR41). FFA3 is expressed in adipose tissue, gut, and the peripheral nervous system and normally activated by short chain fatty acids (SCFAs) produced in the body through the fermentation of complex carbohydrates by the gut microbiota. There is increasing interest in the role of the various free fatty acid receptors and SCFAs as chemical messengers in energy regulation and regulation of inflammatory processes and as therapeutic targets in the treatment of various metabolic and inflammatory conditions. AR420626 has an EC50 value of 117 nM for FFAR3 and did not activate FFAR2 in concentrations up to 100 μM.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Mirelle T A Kleuskens et al.
Allergy, 77(5), 1510-1521 (2021-08-31)
Eosinophilic esophagitis (EoE) is a food allergen driven disease that is accompanied by interleukin (IL) 13 overexpression and esophageal barrier dysfunction allowing transepithelial food allergen permeation. Nutraceuticals, such as short-chain fatty acids (SCFAs) that restore barrier function and increase immune
Yan-Fang Hou et al.
Microbiome, 9(1), 34-34 (2021-02-02)
Parkinson's disease (PD) is a neurodegenerative disorder with no absolute cure. The evidence of the involvement of gut microbiota in PD pathogenesis suggests the need to identify certain molecule(s) derived from the gut microbiota, which has the potential to manage

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service