Skip to Content
Merck
All Photos(1)

Documents

SML1198

Sigma-Aldrich

Terutroban

≥95% (HPLC)

Synonym(s):

(6R)-6-[[(4-Chlorophenyl)sulfonyl]amino]-5,6,7,8-tetrahydro-2-methyl-1-Naphthalenepropanoic acid, (R)-6-[[(4-Chlorophenyl)sulfonyl]amino]-5,6,7,8-tetrahydro-2-methyl-1-Naphthalenepropanoic acid, S18886

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22ClNO4S
CAS Number:
Molecular Weight:
407.91
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

optical activity

[α]/D 25 to 31°, c = 1 in DMSO

storage condition

desiccated

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

O=S(N[C@@H]1CC2=CC=C(C)C(CCC(O)=O)=C2CC1)(C3=CC=C(Cl)C=C3)=O

InChI

1S/C20H22ClNO4S/c1-13-2-3-14-12-16(6-9-19(14)18(13)10-11-20(23)24)22-27(25,26)17-7-4-15(21)5-8-17/h2-5,7-8,16,22H,6,9-12H2,1H3,(H,23,24)/t16-/m0/s1

InChI key

HWEOXFSBSQIWSY-INIZCTEOSA-N

Biochem/physiol Actions

Terutroban is a highly selective and long-acting thromboxane/prostaglandin endoperoxide receptor (TPr) antagonist. The compound Terutroban blocks thromboxane induced platelet aggregation and vasoconstriction. Terutroban prevents vascular wall proliferation and atherogenesis, increases anti-oxidant enzymes like glutathione peroxidase, and has anti-inflammatory properties.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service