Skip to Content
Merck
All Photos(1)

Key Documents

SML0929

Sigma-Aldrich

HS38

≥95% (HPLC)

Synonym(s):

2-[[1-(3-Chlorophenyl)-4,5-dihydro-4-oxo-1H-pyrazolo[3,4-d]pyrimidin-6-yl]thio]-propanamide, 2-[[1-(3-Chlorophenyl)-4-oxo-5H-pyrazolo[5,4-d]pyrimidin-6-yl]sulfanyl]propanamide

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
R$368.00

R$368.00


Estimated to ship onApril 14, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
R$368.00

About This Item

Empirical Formula (Hill Notation):
C14H12ClN5O2S
CAS Number:
Molecular Weight:
349.80
UNSPSC Code:
12352200
NACRES:
NA.77

R$368.00


Estimated to ship onApril 14, 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

Biochem/physiol Actions

HS38 is a potent and selective ATP-competitive inhibitor of Death-associated protein kinases DAPK1 (IC50 = 200 nM, 89% inhibition at 10 RM) and ZIPK (Zipper-interacting protein kinase, also called DAPK3) that regulate programmed cell death, and also inhibits the closely related PIM3 kinase (IC50 = 200 nM, 76% inhibition at 10 RM). In addition to mediating apoptosis, DAPK1 and ZIPK regulate both non-muscle and smooth muscle myosin phosphorylation. HS38 specifically targeted ZIPK in smooth muscle, and decreased contractile force generated in mouse aorta, rabbit ileum, and calyculin A stimulated arterial muscle.
HS38 is a potent and selective ATP-competitive inhibitor that also targets the closely related PIM3 kinase.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Guo et al.
Molecular neurobiology, 56(5), 3368-3379 (2018-08-19)
Loss of memory is an inevitable clinic sign in aging, but its underlying mechanisms remain unclear. Here we show that death-associated protein kinase (DAPK1) is involved in the decays of learning and memory in aging via degradation of Caytaxin, a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service