Skip to Content
Merck
All Photos(1)

Key Documents

SML0876

Sigma-Aldrich

Filastatin

≥98% (HPLC)

Synonym(s):

(3-Chloro-4-methylphenyl)[4-(4-nitrophenyl)-1-piperazinyl]-methanone

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
R$1,834.00

R$1,834.00


Estimated to ship onApril 28, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
R$1,834.00

About This Item

Empirical Formula (Hill Notation):
C18H18ClN3O3
CAS Number:
Molecular Weight:
359.81
UNSPSC Code:
51111800
NACRES:
NA.77

R$1,834.00


Estimated to ship onApril 28, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

2-8°C

SMILES string

Clc1c(ccc(c1)C(=O)N2CCN(CC2)c3ccc(cc3)[N+](=O)[O-])C

InChI

1S/C18H18ClN3O3/c1-13-2-3-14(12-17(13)19)18(23)21-10-8-20(9-11-21)15-4-6-16(7-5-15)22(24)25/h2-7,12H,8-11H2,1H3

InChI key

PNECWWUOUHGWQG-UHFFFAOYSA-N

Biochem/physiol Actions

Filastatin blocks the ability of Candida albicans, and other Candida sp. to bind to polystyrene, and human A549 cell monolayers. Filastatin also blocks biofilm formation and yeast-to-hyphal morphologic transition through inhibition of the hyphal-specific promoter HWP1.
Filastatin is an inhibitor of Candida albicans surface binding, morphology transition and biofilm formation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service