Skip to Content
Merck
All Photos(1)

Key Documents

SML0856

Sigma-Aldrich

AT-56

≥98% (HPLC)

Synonym(s):

4-Dibenzo[a,d]cyclohepten-5-ylidene-1-[4-(2H-tetrazol-5-yl)-butyl]-piperidine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$505.00
25 MG
R$2,038.00

R$505.00


Check Cart for Availability


Select a Size

Change View
5 MG
R$505.00
25 MG
R$2,038.00

About This Item

Empirical Formula (Hill Notation):
C25H27N5
CAS Number:
Molecular Weight:
397.52
UNSPSC Code:
41106300
NACRES:
NA.77

R$505.00


Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

[nH]1nnnc1CCCCN2CCC(=C3c4c(cccc4)C=Cc5c3cccc5)CC2

InChI

1S/C25H27N5/c1-3-9-22-19(7-1)12-13-20-8-2-4-10-23(20)25(22)21-14-17-30(18-15-21)16-6-5-11-24-26-28-29-27-24/h1-4,7-10,12-13H,5-6,11,14-18H2,(H,26,27,28,29)

InChI key

LQNGMDUIRLSESZ-UHFFFAOYSA-N

Biochem/physiol Actions

AT-56 is a selective orally active inhibitor of lipocalin-type prostaglandin D synthase (L-PGDS), one of two synthases involved in the production of Prostaglandin D2 (PGD2) from arachidonic acid. PGD2 is a lipid signaling molecule, which activates two receptors, DP1 involved in centrally mediated processes such as sleep and pain, and DP2 involved and inflammation. The two PGD synthases involved in its synthesis are hematopoietic (H-PDGS) and lipocalin-type (L-PGDS), which acts to form PGD2 in the CNS and other systems but not in inflammatory cells/tissues. AT-56 binds competitively at the enzyme′s catalytic pocket, and has no effect on the production of other PGs or H-PDGS-catalyzed PDG2. AT-56 inhibited PGD2 production by L-PGDS-expressing human TE-671 cells with an IC50 value of 3 μM without affecting production of PGE2 and PGF2, but had no effect on the PGD2 production by H-PGDS-expressing human megakaryocytes.
AT-56 is a selective orally active inhibitor of lipocalin-type prostaglandin D synthase (L-PGDS).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Discover Bioactive Small Molecules for Lipid Signaling Research

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service