Skip to Content
Merck
All Photos(1)

Key Documents

SML0851

Sigma-Aldrich

GZ-793A

≥98% (HPLC)

Synonym(s):

(R)-3-[2,6-cis-Di(4-methoxyphenethyl)piperidin-1-yl]propane-1,2-diol hydrochloride, 3-[(2R,6S)-2,6-Bis[2-(4-methoxyphenyl)ethyl]-1-piperidinyl]-(2R)-1,2-propanediol hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
R$1,831.00

R$1,831.00


Estimated to ship onMay 31, 2025



Select a Size

Change View
25 MG
R$1,831.00

About This Item

Empirical Formula (Hill Notation):
C26H37NO4 · HCl
CAS Number:
Molecular Weight:
464.04
UNSPSC Code:
12352200
NACRES:
NA.77

R$1,831.00


Estimated to ship onMay 31, 2025


Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 3 mg/mL, clear

storage temp.

2-8°C

SMILES string

Cl.N1([C@H](CCC[C@H]1CCc3ccc(cc3)OC)CCc2ccc(cc2)OC)C[C@@H](O)CO

InChI

1S/C26H37NO4.ClH/c1-30-25-14-8-20(9-15-25)6-12-22-4-3-5-23(27(22)18-24(29)19-28)13-7-21-10-16-26(31-2)17-11-21;/h8-11,14-17,22-24,28-29H,3-7,12-13,18-19H2,1-2H3;1H/t22-,23+,24-;/m1./s1

InChI key

FLFFCQKGCYZDOI-SJUVJREQSA-N

Biochem/physiol Actions

GZ-793A is a selective inhibitor of vesicular monoamine transporter 2 (VMAT2), which transports the monoamine neurotransmitters from cellular cytosol into synaptic vesicles. GZ-793A is 50-fold selective for VMAT over the serotonin or dopamine transporters. VMAT2 has recently been considered a target for the treatment of methamphetamine abuse. In rat studies, GZ-793A decreased self-administration of methamphetamine with no development of tolerance.
GZ-793A is a selective inhibitor of vesicular monoamine transporter 2 (VMAT2).

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shunsuke Kimura et al.
PloS one, 16(1), e0245526-e0245526 (2021-01-20)
Neuroblastoma, the most common extracranial solid malignancy among children, originates from undifferentiated neural crest cells (NCC). Despite recent intensified treatment, high-risk patients still have a high mortality rate. To explore a new therapeutic strategy, we performed an integrated genomic and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service