Skip to Content
Merck
All Photos(1)

Documents

SML0845

Sigma-Aldrich

DAU-5884

≥98% (HPLC)

Synonym(s):

8-Methyl-8-azabicyclo-3-endo[3.2.1]oct-3-yl 1,4-dihydro-2-oxo-3(2H)-quinazolinecarboxylic acid ester, 8-Methyl-8-azabicyclo[3.2.1]oct-3-yl ester 1,4-dihydro-2-oxo-3(2H)-quinazolinecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21N3O3
CAS Number:
Molecular Weight:
315.37
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

−20°C

SMILES string

CN1[C@]2([H])CC[C@@]1([H])C[C@@H](OC(N3CC4=CC=CC=C4NC3=O)=O)C2

InChI

1S/C17H21N3O3/c1-19-12-6-7-13(19)9-14(8-12)23-17(22)20-10-11-4-2-3-5-15(11)18-16(20)21/h2-5,12-14H,6-10H2,1H3,(H,18,21)/t12-,13+,14?

InChI key

NWXCDQHBVVPPTL-PBWFPOADSA-N

Biochem/physiol Actions

DAU-5884 is a M3-selective muscarinic receptor antagonist. The compound DAU-5884 blocks cobrotoxin-induced transient A-type K(+) currents in dorsal root ganglion neurons.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshihiro Mitoh et al.
Autonomic neuroscience : basic & clinical, 206, 1-7 (2017-06-11)
The superior salivatory nucleus (SSN) contains parasympathetic preganglionic neurons innervating the submandibular and sublingual salivary glands. Cevimeline, a muscarinic acetylcholine receptor (mAChR) agonist, is a sialogogue that possibly stimulates SSN neurons in addition to the salivary glands themselves because it

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service