Skip to Content
Merck
All Photos(1)

Documents

SML0827

Sigma-Aldrich

FPH1

≥98% (HPLC)

Synonym(s):

2-(N-(5-Chloro-2-methylphenyl)(methylsulfonamido)-N-(2,6-difluorophenyl)-acetamide, 2-[(5-Chloro-2-methylphenyl)(methylsulfonyl)amino]-N-(2,6-difluorophenyl)-acetamide, BRD-6125

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15ClF2N2O3S
CAS Number:
Molecular Weight:
388.82
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

InChI

1S/C16H15ClF2N2O3S/c1-10-6-7-11(17)8-14(10)21(25(2,23)24)9-15(22)20-16-12(18)4-3-5-13(16)19/h3-8H,9H2,1-2H3,(H,20,22)

InChI key

WAOBCCBUTHNTFO-UHFFFAOYSA-N

Biochem/physiol Actions

FPH1 (BRD-6125) induces functional proliferation of primary human hepatocytes in vitro. Human induced pluripotent stem (iPS) cells can be differentiated toward hepatocyte-like (iHep) cells, but replication is not great, and cells typically show an immature hepatic phenotype and have limited use as a renewable source of functional human hepatocytes. FPH1 induced hepatocyte colonies were used in conjunction with FH1 (BRD-K4477) to induce a more mature phenotype. Cultures treated with both FH1 and FPH1 contained larger colonies of hepatocyte-like (iHep) cells, which showed more pronounced hepatocyte morphologies with a more mature phenotype than previously obtainable.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service