Skip to Content
Merck
All Photos(1)

Key Documents

SML0813

Sigma-Aldrich

BRD56491

≥98% (HPLC)

Synonym(s):

2-(4-Methylphenyl)-4H-benzo[h]chromen-4-one, 2-(4-Methylphenyl)-4H-naphtho[1,2-b]pyran-4-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$563.00
25 MG
R$2,270.00

R$563.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
R$563.00
25 MG
R$2,270.00

About This Item

Empirical Formula (Hill Notation):
C20H14O2
CAS Number:
Molecular Weight:
286.32
UNSPSC Code:
12352200
NACRES:
NA.77

R$563.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

Biochem/physiol Actions

BRD56491 is a nontoxic enhancer of reactive oxygen species (ROS) that strongly elevates markers of oxidative stress without causing cell death.
BRD56491 is a nontoxic enhancer of reactive oxygen species (ROS) that strongly elevates markers of oxidative stress without causing cell death. However, combining BRD56491 with nontoxic doses of the glutathione synthesis inhibitor L-buthionine sulfoximine did lead to cell death in a variety of cancer cell lines. BRD56491 can be used to create a more oxidizing cell state and aid in studies of cancer cell lines resilient to induced increases in ROS levels.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service