Skip to Content
Merck
All Photos(1)

Key Documents

SML0759

Sigma-Aldrich

L002

≥98% (HPLC)

Synonym(s):

4-[O-[(4-Methoxyphenyl)sulfonyl]oxime]-2,6-dimethyl-2,5-cyclohexadiene-1,4-dione, NSC764414

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H15NO5S
CAS Number:
Molecular Weight:
321.35
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(ON=C2C=C(C(=O)C(=C2)C)C)c1ccc(cc1)OC

InChI

1S/C15H15NO5S/c1-10-8-12(9-11(2)15(10)17)16-21-22(18,19)14-6-4-13(20-3)5-7-14/h4-9H,1-3H3

InChI key

VEWFTYOFWIXCIO-UHFFFAOYSA-N

Biochem/physiol Actions

L002 (NSC764414) is an inhibitor of acetyltransferase p300, a lysine acetyltransferase that catalyzes acetyl group attachment to lysine residues of a variety of proteins including histones and p53, and acts as a critical coactivator of several oncogenic transcription factors involved in cancer cell survival and proliferation including STAT3, NF-kB, and hypoxia-inducible factor-1α (HIF-1α). L002 (NSC764414) was discovered from compounds assayed for cytotoxicity to the triple-negative breast cancer (TNBC) cell line MDA-MB-231 but not to the human mammary epithelial cells, then further screened for inhibition of p300. L002 has an in vitro IC50 of 1.98 μM for p300 with lesser inhibition of the GNAT (GCN5-related N-acetyltransferase) family, and no inhibition against the MYST family of histone acetyltransferases (HATs), histone deacetylases (HDACs), or histone methyltransferases (HMTs). L002 potently suppressed tumor growth and histone acetylation of MDA-MB-468 xenografts and leukemia and lymphoma cell lines.
L002 (NSC764414) is an inhibitor of acetyltransferase p300.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service