Skip to Content
Merck
All Photos(1)

Key Documents

SML0745

Sigma-Aldrich

NS6180

≥98% (HPLC)

Synonym(s):

4-[[3-(Trifluoromethyl)phenyl]methyl]-2H-1,4-benzothiazin-3(4H)-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$519.00

R$519.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
R$519.00

About This Item

Empirical Formula (Hill Notation):
C16H12F3NOS
CAS Number:
Molecular Weight:
323.33
UNSPSC Code:
12352200
NACRES:
NA.77

R$519.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC(F)(F)c1cc(ccc1)CN2c3c(cccc3)SCC2=O

InChI

1S/C16H12F3NOS/c17-16(18,19)12-5-3-4-11(8-12)9-20-13-6-1-2-7-14(13)22-10-15(20)21/h1-8H,9-10H2

InChI key

ZUIJXKLTUFCDGO-UHFFFAOYSA-N

Biochem/physiol Actions

NS6180 is a novel inhibitor of the intermediate-conductance Ca2+-activated K+ channel (KCa3.1) The IC50 of NS6180 against cloned human KCa3.1 is 9 nM. The compound blocks proliferation and production of IL-2 and IFNγ with sub-micromolar potencies in either ConA or PMA/ionomycin challenged rat splenocytes.
NS6180 is a novel inhibitor of the intermediate-conductance Ca2+-activated K+ channel (KCa3.1).

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to potassium channels for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service