Skip to Content
Merck
All Photos(1)

Key Documents

SML0723

Sigma-Aldrich

ML252

≥98% (HPLC)

Synonym(s):

(S)-2-Phenyl-N-(2-(pyrrolidin-1-yl)phenyl)butanamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$611.00
25 MG
R$2,467.00

R$611.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
R$611.00
25 MG
R$2,467.00

About This Item

Empirical Formula (Hill Notation):
C20H24N2O
CAS Number:
Molecular Weight:
308.42
UNSPSC Code:
12352200
NACRES:
NA.77

R$611.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

, light to dark purple

solubility

DMSO: 25 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

ML252 is a potent, brain penetrant potassium channel Kv7.2 (KCNQ2) inhibitor with an IC50 of 69 nM. ML252 is only slightly selective over KCNQ3 and KCNQ4 but is over 40-fold selective for KCNQ2 vs KCNQ1, unlike other KCNQ inbhibitors currently used. ML252 was also selective vs. >68 tested GPCRs, ion channels, and transporters. Potassium channel Kv7.2 has become a new target for Alzheimer′s Disease drug research because inhibiting it enhances acetylcholine release. SAR studies showed a small structural change from ethyl group to hydrogen resulted in a functional shift from antagonist to agonist activity (37, EC50 of 170 nM), suggesting an interaction at a critical site for controlling KCNQ2 channel gating.
ML252 is a potent, brain penetrant potassium channel Kv7.2 (KCNQ2) inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keyong Li et al.
Cell reports, 34(5), 108714-108714 (2021-02-04)
Brainstem networks that control regular tidal breathing depend on excitatory drive, including from tonically active, CO2/H+-sensitive neurons of the retrotrapezoid nucleus (RTN). Here, we examine intrinsic ionic mechanisms underlying the metronomic firing activity characteristic of RTN neurons. In mouse brainstem

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service