Skip to Content
Merck
All Photos(1)

Key Documents

SML0633

Sigma-Aldrich

UNC0646

≥98% (HPLC)

Synonym(s):

N-(1-Cyclohexyl-4-piperidinyl)-2-[hexahydro-4-(1-methylethyl)-1H-1,4-diazepin-1-yl]-6-methoxy-7-[3-(1-piperidinyl)propoxy]-4-quinazolinamine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$654.00
25 MG
R$2,621.00

R$654.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
R$654.00
25 MG
R$2,621.00

About This Item

Empirical Formula (Hill Notation):
C36H59N7O2
CAS Number:
Molecular Weight:
621.90
UNSPSC Code:
12352200
NACRES:
NA.77

R$654.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

N2(CCC(CC2)Nc3nc(nc5c3cc(c(c5)OCCCN6CCCCC6)OC)N4CCN(CCC4)C(C)C)C1CCCCC1

InChI

1S/C36H59N7O2/c1-28(2)41-19-10-20-43(24-23-41)36-38-32-27-34(45-25-11-18-40-16-8-5-9-17-40)33(44-3)26-31(32)35(39-36)37-29-14-21-42(22-15-29)30-12-6-4-7-13-30/h26-30H,4-25H2,1-3H3,(H,37,38,39)

InChI key

OUKWLRHRXOPODD-UHFFFAOYSA-N

Biochem/physiol Actions

UNC0646 is potent, selective inhibitor of the methyltransferase G9a, which catalyzes lysine-methylation of histones, and nonhistone proteins including p53.
UNC0646 is potent, selective inhibitor of the methyltransferase G9a.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David A Vierra et al.
Oncotarget, 8(44), 76443-76457 (2017-11-05)
Fanconi anemia (FA) is a rare disease characterized by congenital defects, bone marrow failure, and atypically early-onset cancers. The FA proteins function cooperatively to repair DNA interstrand crosslinks. A major step in the activation of the pathway is the monoubiquitination

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service