Skip to Content
Merck
All Photos(1)

Key Documents

SML0617

Sigma-Aldrich

A-804598

≥98% (HPLC)

Synonym(s):

N-Cyano-N′′-[(1S)-1-phenylethyl]-N′-5-quinolinyl-guanidine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$566.00
25 MG
R$2,276.00

R$566.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
R$566.00
25 MG
R$2,276.00

About This Item

Empirical Formula (Hill Notation):
C19H17N5
CAS Number:
Molecular Weight:
315.37
UNSPSC Code:
12352200
NACRES:
NA.77

R$566.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

N(c2c3c(nccc3)ccc2)\C(=N/[C@@H](C)c1ccccc1)\NC#N

InChI

1S/C19H17N5/c1-14(15-7-3-2-4-8-15)23-19(22-13-20)24-18-11-5-10-17-16(18)9-6-12-21-17/h2-12,14H,1H3,(H2,22,23,24)/t14-/m0/s1

InChI key

PQYCRDPLPKGSME-AWEZNQCLSA-N

Biochem/physiol Actions

A-804598 is a P2X7 selective, competitive antagonist.
A-804598 is a P2X7 selective, competitive antagonist. In competition assays, the IC50s for human, rat and mouse channels are 11, 10 and 9 nM, respectively.

Features and Benefits

This compound is featured on the P2 Receptors: P2X Ion Channel Family page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to P2X ion channel family receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service