Skip to Content
Merck
All Photos(1)

Key Documents

SML0537

Sigma-Aldrich

MI-1

≥98% (HPLC)

Synonym(s):

4-[4-(4,5-Dihydro-5,5-dimethyl-2-thiazolyl)-1-piperazinyl]-5,6,7,8-tetrahydro-[1]benzothieno[2,3-d]pyrimidine, CCG21397

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H25N5S2
CAS Number:
Molecular Weight:
387.57
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL (clear solution, warmed)

storage temp.

2-8°C

SMILES string

CC1(C)CN=C(S1)N2CCN(CC2)c3ncnc4sc5CCCCc5c34

InChI

1S/C19H25N5S2/c1-19(2)11-20-18(26-19)24-9-7-23(8-10-24)16-15-13-5-3-4-6-14(13)25-17(15)22-12-21-16/h12H,3-11H2,1-2H3

InChI key

MNSXQGKLDTVOLP-UHFFFAOYSA-N

Biochem/physiol Actions

MI-1 is a Menin-MLL interaction inhibitor. The mixed lineage leukemia (MLL) gene encodes MLL, a histone methyltransferase important in regulating gene transcription. MLL rearrangements and formation of fusion oncoproteins can block hematopoietic differentiation and increase proliferation, resulting in acute leukemias. Menin, encoded by the MEN1 gene, is a tumor suppressor associated with multiple endocrine neoplasia type 1. It also acts as an oncogenic co-factor of MLL fusion proteins. MI-1 inhibits Menin-MLL interaction with an IC50 of 1.9 micromolar.
MI-1is a Menin-MLL interaction inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sheng Cao et al.
Journal of cardiovascular pharmacology, 70(1), 25-33 (2017-07-06)
Intravenous ultrasound-targeted microbubble destruction (IV-UTMD) has made distinct but limited progress in gene therapy. Intracoronary (IC) injection may lead to more gene transfection than IV injection. This study compared the therapeutic effects of IC-UTMD-mediated and conventional IV-UTMD-mediated gene transfection in
Amanda N Shelton et al.
The ISME journal, 13(3), 789-804 (2018-11-16)
The vitamin B12 family of cofactors known as cobamides are essential for a variety of microbial metabolisms. We used comparative genomics of 11,000 bacterial species to analyze the extent and distribution of cobamide production and use across bacteria. We find

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service