Skip to Content
Merck
All Photos(1)

Key Documents

SML0384

Sigma-Aldrich

Eicosapentaenoyl ethanolamide

5 mg/mL in absolute ethanol, ≥98% (HPLC)

Synonym(s):

(5Z,8Z,11Z,14Z,17Z)-N-(2-Hydroxyethyl)-5,8,11,14,17-eicosapentaenamide, EPEA

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$429.00

R$429.00


Check Cart for Availability


Select a Size

Change View
5 MG
R$429.00

About This Item

Empirical Formula (Hill Notation):
C22H35NO2
CAS Number:
Molecular Weight:
345.52
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

R$429.00


Check Cart for Availability

Assay

≥98% (HPLC)

form

solution

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

storage condition

protect from light
under inert gas

concentration

5 mg/mL in absolute ethanol

color

light yellow

storage temp.

−20°C

SMILES string

CC\C=C/C\C=C/C\C=C/C\C=C/C\C=C/CCCC(=O)NCCO

InChI

1S/C22H35NO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-22(25)23-20-21-24/h3-4,6-7,9-10,12-13,15-16,24H,2,5,8,11,14,17-21H2,1H3,(H,23,25)/b4-3-,7-6-,10-9-,13-12-,16-15-

InChI key

OVKKNJPJQKTXIT-JLNKQSITSA-N

Biochem/physiol Actions

Eicosapentaenoyl ethanolamide (EPEA) is a lipid mediator that has been found to suppress lifespan extension resulting from dietary restriction in C. elegans and also to have antiproliferative and anti-inflammatory actions. In C. elegans, it is believed to act as a metabolic signal that couples nutrient availability with growth and lifespan. EPEA is a member of the N-acylethanolamines (NAEs), lipid-derived signaling molecules that include the mammalian endocannabinoid arachidonoyl ethanolamide (AEA). In mammals, NAEs are believed to act primarily through cannabinoid receptors, although they can also interact with a variety of other targets, and EPEA has been shown to act as a CB1 and CB2 receptor agonist. However, C. elegans does not possess clear orthologues of the mammalian cannabinoid receptors which suggests that there are unidentified NAE receptors in nematodes that are possibly conserved mediators of NAE signaling.
Eicosapentaenoyl ethanolamide (EPEA) is a lipid mediator; suppresses lifespan extension; anti proliferative and anti-inflammatory.

Other Notes

Light and air sensitive; store in a freezer.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Strahil Iv Pastuhov et al.
Nature communications, 3, 1136-1136 (2012-10-18)
The ability of neurons to regenerate their axons after injury is determined by a balance between cellular pathways that promote and those that inhibit regeneration. In Caenorhabditis elegans, axon regeneration is positively regulated by the c-Jun N-terminal kinase mitogen activated
Michiel G J Balvers et al.
Metabolomics : Official journal of the Metabolomic Society, 8(6), 1130-1147 (2012-11-09)
It is well established that dietary intake of n-3 fatty acids is associated with anti-inflammatory effects, and this has been linked to modulation of the oxylipin and endocannabinoid metabolomes. However, the amount of data on specific tissue effects is limited
Mark Lucanic et al.
Nature, 473(7346), 226-229 (2011-05-13)
Dietary restriction is a robust means of extending adult lifespan and postponing age-related disease in many species, including yeast, nematode worms, flies and rodents. Studies of the genetic requirements for lifespan extension by dietary restriction in the nematode Caenorhabditis elegans
Tobias Dietsche et al.
PLoS pathogens, 12(12), e1006071-e1006071 (2016-12-16)
Bacterial type III protein secretion systems inject effector proteins into eukaryotic host cells in order to promote survival and colonization of Gram-negative pathogens and symbionts. Secretion across the bacterial cell envelope and injection into host cells is facilitated by a
Gil Bar-Sela et al.
Cancers, 12(9) (2020-09-03)
Cannabis or its derivatives are widely used by patients with cancer to help with cancer symptoms and treatment side effects. However, cannabis has potent immunomodulatory properties. To determine if cannabis consumption during immunotherapy affects therapy outcomes, we conducted a prospective

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service