Skip to Content
Merck
All Photos(1)

Key Documents

SML0381

Sigma-Aldrich

ALLO-1

≥98% (HPLC)

Synonym(s):

3-(4-Chlorophenyl)-5-methyl-1-(phenylmethyl)-2,4-Imidazolidinedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H15ClN2O2
CAS Number:
Molecular Weight:
314.77
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

CC1N(Cc2ccccc2)C(=O)N(C1=O)c3ccc(Cl)cc3

InChI

1S/C17H15ClN2O2/c1-12-16(21)20(15-9-7-14(18)8-10-15)17(22)19(12)11-13-5-3-2-4-6-13/h2-10,12H,11H2,1H3

InChI key

GHAJNZREWTZYQJ-UHFFFAOYSA-N

Biochem/physiol Actions

ALLO-1 is a Smoothened (Smo) antagonist that inhibits both wild-type and mutant Smo, including D473H SMO mutant. ALLO-1 binds to a different domain of Smo than other known Smo ligands.
ALLO-1 is a Smoothened (Smo) antagonist that inhibits both wild-type and mutant Smo.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miyuki Sato et al.
Nature cell biology, 20(1), 81-91 (2017-12-20)
In Caenorhabditis elegans embryos, paternally provided organelles, including mitochondria, are eliminated by a process of selective autophagy called allophagy, the mechanism by which mitochondrial DNA is inherited maternally. However, it remains unclear how paternal organelles are recognized and targeted for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service