Skip to Content
Merck
All Photos(1)

Key Documents

SML0342

Sigma-Aldrich

TFM-4AS-1

≥98% (HPLC)

Synonym(s):

(4aR,6aS,7S,11aR)-1,4a,6a-Trimethyl-2-oxo-N-[2-(trifluoromethyl)phenyl]-2,4a,4b,5,6,6a,7,8,9,9a,9b,10,11,11atetradecahydro-1H-indeno[5,4-f]quinoline-7-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$1,123.00
25 MG
R$5,093.00

R$1,123.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
R$1,123.00
25 MG
R$5,093.00

About This Item

Empirical Formula (Hill Notation):
C27H33F3N2O2
CAS Number:
Molecular Weight:
474.56
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

R$1,123.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: >5 mg/mL

storage temp.

2-8°C

SMILES string

CN1[C@@H]2CC[C@@H]3[C@@H]4CC[C@H](C(=O)Nc5ccccc5C(F)(F)F)[C@@]4(C)CC[C@@H]3[C@@]2(C)C=CC1=O

InChI

1S/C27H33F3N2O2/c1-25-14-12-18-16(8-11-22-26(18,2)15-13-23(33)32(22)3)17(25)9-10-20(25)24(34)31-21-7-5-4-6-19(21)27(28,29)30/h4-7,13,15-18,20,22H,8-12,14H2,1-3H3,(H,31,34)/t16-,17+,18+,20-,22-,25+,26-/m1/s1

InChI key

YFBLEKKYWFJKBP-HNXMFXTHSA-N

Related Categories

Biochem/physiol Actions

TFM-4AS-1 is a potent SARM (Selective Androgen Receptor Modulator) and partial agonist of AR.
TFM-4AS-1 is a potent SARM (Selective Androgen Receptor Modulator). TFM-4AS-1 is a partial agonist of AR. The compound promotes the buildup of bone and muscle mass while having reduced effects on reproductive organs and sebaceous glands. The compound also is an inhibitor of rat 5 α-reductase enzymes type I (2 nM) and type 2 (3 nM).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service