Skip to Content
Merck
All Photos(1)

Key Documents

SML0258

Sigma-Aldrich

CPPG

≥98% (HPLC)

Synonym(s):

(R,S)-α-Cyclopropyl-4-phosphonophenylglycine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14NO5P
CAS Number:
Molecular Weight:
271.21
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: >5 mg/mL

storage temp.

room temp

SMILES string

NC(C1CC1)(C(O)=O)c2ccc(cc2)P(O)(O)=O

InChI

1S/C11H14NO5P/c12-11(10(13)14,7-1-2-7)8-3-5-9(6-4-8)18(15,16)17/h3-7H,1-2,12H2,(H,13,14)(H2,15,16,17)

InChI key

IGODGTDUQSMDQU-UHFFFAOYSA-N

Biochem/physiol Actions

(R,S)-α-Cyclopropyl-4-phosphonophenylglycine (CPPG) can be used in combination with scopolamine to treat the depressive disorder.
CPPG is a potent group II/III metabotropic glutamate receptor antagonist, with approximately 20-fold selectivity for mGlu group III over group II (IC50 values of 2.2 and 46.2 nM respectively).
CPPG is a potent group II/III metabotropic glutamate receptor antagonist.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kouji Fukuyama et al.
Biomolecules, 9(6) (2019-06-20)
Pharmacological mechanisms of gold-standard antipsychotics against treatment-refractory schizophrenia, such as clozapine (CLZ), remain unclear. We aimed to explore the mechanisms of CLZ by investigating the effects of MK801 and CLZ on tripartite synaptic transmission in the thalamocortical glutamatergic pathway using
Emmanuel Dornier et al.
Nature communications, 8(1), 2255-2255 (2017-12-23)
The role of glutaminolysis in providing metabolites to support tumour growth is well-established, but the involvement of glutamine metabolism in invasive processes is yet to be elucidated. Here we show that normal mammary epithelial cells consume glutamine, but do not
Mohaddeseh Ebrahimi-Ghiri et al.
Physiology & behavior, 224, 113034-113034 (2020-07-01)
Clinical trials have revealed that the muscarinic receptor antagonist scopolamine produces a fast and potent antidepressant response. However, the anticholinergic adverse effects and the risk of psychosis at higher doses limit the widespread clinical use of scopolamine. Combination therapy of
Xing Yang et al.
Frontiers in bioengineering and biotechnology, 8, 903-903 (2020-10-30)
A simple and facile one-step method was developed to construct a small molecular prodrug amphiphile self-assembled organic dots CPPG with aggregation-induced emission (AIE) characteristics. Diphenylalanine peptide (FF), which is the essential moiety of the self-assembling peptide-drug conjugate and as its
Gelareh Abulwerdi et al.
Neurochemistry international, 138, 104770-104770 (2020-05-27)
Chronic dysregulated microglial activation may lead to persistent inflammation and progressive neurodegeneration. A previous study reported that ADX88178, a putative metabotropic glutamate receptor 4 (mGluR4) positive allosteric modulator (PAM), exerts anti-inflammatory effects in microglia by activating mGluR4. We employed in

Articles

Sigma-Aldrich offers many products related to G-protein family glutamate receptors for your research needs.

DISCOVER Bioactive Small Molecules for Neuroscience

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service