Skip to Content
Merck
All Photos(1)

Documents

SML0257

Sigma-Aldrich

JZL195

≥98% (HPLC)

Synonym(s):

4-[(3-Phenoxyphenyl)methyl]-1-piperazinecarboxylic acid 4-nitrophenyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H23N3O5
CAS Number:
Molecular Weight:
433.46
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: ≥5 mg/mL at warmed

storage temp.

−20°C

SMILES string

[O-][N+](=O)c1ccc(OC(=O)N2CCN(CC2)Cc3cccc(Oc4ccccc4)c3)cc1

InChI

1S/C24H23N3O5/c28-24(32-22-11-9-20(10-12-22)27(29)30)26-15-13-25(14-16-26)18-19-5-4-8-23(17-19)31-21-6-2-1-3-7-21/h1-12,17H,13-16,18H2

InChI key

QNYRAEKLMNDRFY-UHFFFAOYSA-N

Application

JZL195 has been used:
  • as a selective inhibitor of endocannabinoid (eCB) clearance enzymes to induce in vivo long-term depression at CA3-CA1 synapses and at prelimbic (PrL)-nucleus accumbens (NAc)synapses, to study the neuroprotective action of eCB
  • to inhibit the action of hydrolytic enzymes that limit eCB activity, to study the effect of 2-linoleoylglycerol (2-LG) on the human CB1 receptor activity
  • as a dual fatty acid amide hydrolase (FAAH)/monoacylglycerol lipase (MAGL) inhibitor to study its dose-related antipruritic effect on the serotonin (5-HT)-induced scratching model

Biochem/physiol Actions

JZL195 is a potent dual inhibitor of Monoacylglycerol lipase (MAGL) and fatty acid amide hydrolase (FAAH), enzymes that degrade the endocannabinoids 2-arachidonoylglycerol (2-AG) and anandamide (AEA), the endogenous ligands for the cannabinoid G-protein coupled receptors CB1 and CB2. IC50 values are 2 nM for MAGL and 4 nM for FAAH. JZL195 has been shown to inhibit endocannabinoid hydrolysis and elevate 2-AG and AEA levels in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Feng Wang et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 39(6), 1122-1137 (2018-02-13)
Ischemia not only activates cell death pathways but also triggers endogenous protective mechanisms. However, it is largely unknown what is the essence of the endogenous neuroprotective mechanisms induced by preconditioning. In this study we demonstrated that systemic injection of JZL195
Ozgur Yesilyurt et al.
Archives of dermatological research, 308(5), 335-345 (2016-04-30)
The increase of endocannabinoid tonus by inhibiting fatty acid amide hydrolase (FAAH) or monoacylglycerol lipase (MAGL) represents a promising therapeutic approach in a variety of disease to overcome serious central side effects of exocannabinoids. Recent studies reported that systemic administration
Tania Muller et al.
American journal of physiology. Endocrinology and metabolism, 313(1), E26-E36 (2017-03-23)
Evidence has accumulated that obesity-related metabolic dysregulation is associated with overactivation of the endocannabinoid system (ECS), which involves cannabinoid receptor 1 (CB1R), in peripheral tissues, including adipose tissue (AT). The functional consequences of CB1R activation on AT metabolism remain unclear.
Leanne Lu et al.
Cannabis and cannabinoid research, 4(4), 231-239 (2019-12-25)
Introduction: The cannabinoid type 1 (CB1) receptor and cannabinoid type 2 (CB2) receptor are widely expressed in the body and anandamide (AEA) and 2-arachidonoylglycerol (2-AG) are their best characterized endogenous ligands. The diacylglycerol lipases (diacylglycerol lipase alpha and diacylglycerol lipase
Shivani Jaiswal et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 107, 1611-1623 (2018-09-28)
Fatty acid amide hydrolase (FAAH) represents a potential therapeutic target for number of peripheral and nervous system related disorders including neuropathic pain and neuroinflammation. A library of N-(2,4-dichlorobenzoyl) isatin Schiff bases 7a-7l and 8a-8c were designed using the contemporary scaffold-hopping

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service