Skip to Content
Merck
All Photos(1)

Key Documents

SML0066

Sigma-Aldrich

NGB 2904 hydrochloride

≥98% (HPLC)

Synonym(s):

N-[4-[4-(2,3-Dichlorophenyl)-1-piperazinyl]butyl]-9H-fluorene-2-carboxamide hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$1,008.00
25 MG
R$4,077.00

R$1,008.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
R$1,008.00
25 MG
R$4,077.00

About This Item

Empirical Formula (Hill Notation):
C28H29Cl2N3O·HCl
CAS Number:
Molecular Weight:
530.92
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

R$1,008.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to tan

solubility

DMSO: ≥25 mg/mL

storage temp.

2-8°C

SMILES string

Cl.Clc1cccc(N2CCN(CCCCNC(=O)c3ccc-4c(Cc5ccccc-45)c3)CC2)c1Cl

InChI

1S/C28H29Cl2N3O.ClH/c29-25-8-5-9-26(27(25)30)33-16-14-32(15-17-33)13-4-3-12-31-28(34)21-10-11-24-22(19-21)18-20-6-1-2-7-23(20)24;/h1-2,5-11,19H,3-4,12-18H2,(H,31,34);1H

InChI key

PFIWYJNBKGCVFM-UHFFFAOYSA-N

Application

NGB 2904 hydrochloride may be used in cell signaling studies.

Biochem/physiol Actions

Inhibition of dopamine D3 receptor function by NGB 2904 increases spontaneous locomotor activity in rodents.[1]
NGB 2904 hydrochloride is a potent and selective dopamine D3 receptor antagonist
NGB 2904 is a potent and selective dopamine D3 receptor antagonist with a D3/D2 ratio of 800 for rat and high binding affinity (1.4 nM) at the hD3 receptor with 150-fold selectivity for primate D3 receptors over primate D2L receptors. It attenuates the rewarding effects of cocaine and methamphetamine.

Features and Benefits

This compound is featured on the Dopamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laurel M Pritchard et al.
Pharmacology, biochemistry, and behavior, 86(4), 718-726 (2007-04-06)
The dopamine D3 receptor is believed to play an important role in regulation of rodent locomotor behavior, and has been proposed as a therapeutic target for substance abuse, psychotic disorders, and Parkinson's disease. One model of dopamine D3 receptor function

Articles

We offer many products related to dopamine receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service