Skip to Content
Merck
All Photos(1)

Key Documents

SML0010

Sigma-Aldrich

LY-334370 hydrochloride hydrate

≥98% (HPLC), powder

Synonym(s):

4-Fluoro-N-[3-(1-methyl-4-piperidinyl)-1H-indol-5-yl]benzamide hydrochloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22FN3O · HCl · xH2O
CAS Number:
Molecular Weight:
387.88 (anhydrous basis)
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

H2O: ≥5 mg/mL (warmed)

originator

Eli Lilly

storage temp.

2-8°C

SMILES string

Cl.CN1CCC(CC1)c2c[nH]c3ccc(NC(=O)c4ccc(F)cc4)cc23

InChI

1S/C21H22FN3O.ClH/c1-25-10-8-14(9-11-25)19-13-23-20-7-6-17(12-18(19)20)24-21(26)15-2-4-16(22)5-3-15;/h2-7,12-14,23H,8-11H2,1H3,(H,24,26);1H

InChI key

DGPDGAPZTPSHBL-UHFFFAOYSA-N

Application

LY-334370 hydrochloride hydrate may be used to study 5-HT 1F-mediated cell signaling.

Biochem/physiol Actions

LY-334370 hydrochloride is a selective 5-HT1F receptor agonist.
LY-334370 modulates the transmission in the mouse trigeminal system and inhibits the neuronal responses within rat Sp5C by inhibiting c-Fos activation. It shows efficacy in aborting migraine attacks.[1][2]

Features and Benefits

This compound was developed by Eli Lilly. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandra A Filla et al.
Journal of medicinal chemistry, 46(14), 3060-3071 (2003-06-27)
Compound 1a (LY334370), a selective 5-HT(1F) receptor agonist (SSOFRA), inhibited dural inflammation in the neurogenic plasma protein extravasation model of migraine and demonstrated clinical efficacy for the acute treatment of migraine. Although 1a was greater than 100-fold selective over both
D D Mitsikostas et al.
Cephalalgia : an international journal of headache, 22(5), 384-394 (2002-07-12)
In order to investigate the c-fos response within the trigeminal nucleus caudalis (Sp5C) after noxious meningeal stimulation, capsaicin (0.25, 0.5, 1 and 5 nmol) was administered intracisternally in urethane (1 g/kg) and alpha-chloralose (20 mg/kg) anaesthetized male mice. Capsaicin induced

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service