Skip to Content
Merck
All Photos(1)

Key Documents

SMB01008

Sigma-Aldrich

Aschantin

≥90% (LC/MS-ELSD)

Synonym(s):

(+)-Aschantin, 5-[(3S,3aR,6S,6aR)-6-(3,4,5-trimethoxyphenyl)-1,3,3a,4,6,6a-hexahydrofuro[3,4-c]furan-3-yl]-1,3-benzodioxole

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
R$3,066.00

R$3,066.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
R$3,066.00

About This Item

Empirical Formula (Hill Notation):
C22H24O7
CAS Number:
Molecular Weight:
400.42
UNSPSC Code:
12352205
NACRES:
NA.25

R$3,066.00


Check Cart for Availability

Request a Bulk Order

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

400.42

solubility

water: slightly soluble

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O1[C@@H]([C@@H]3[C@@H]([C@H](OC3)c4cc5c(cc4)OCO5)C1)c2cc(c(c(c2)OC)OC)OC

InChI

1S/C22H24O7/c1-23-18-7-13(8-19(24-2)22(18)25-3)21-15-10-26-20(14(15)9-27-21)12-4-5-16-17(6-12)29-11-28-16/h4-8,14-15,20-21H,9-11H2,1-3H3/t14-,15-,20+,21+/m0/s1

InChI key

ONDWGDNAFRAXCN-VUEDXXQZSA-N

General description

Aschantin, a bioactive lignan, is a natural product commonly available from Magnolia Sp, and Artemisia Sp. plants. This secondary metabolite derived from plant sources exhibits various biological activities such as anti-inflammatory, antioxidant, and anticancer activities.

Application

It is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Biochem/physiol Actions

Aschantin has various biological activities including peroxynitrite scavenging capacity, inhibition of inducible NO synthetase, antiplasmodial activity, Ca2+-antagonistic activity, platelet activating factor-antagonistic activity, and chemo-preventative or therapeutic activity mediated via inhibition of mTOR kinase.
The anticancer activity of aschantin is achieved by inhibiting Akt, a protein that plays a crucial role in promoting cancer cell growth and proliferation. By blocking Akt, aschantin disrupts multiple signaling pathways that are essential for cancer cells to thrive and multiply. This disruption ultimately leads to the suppression of cancer cell proliferation, making it an effective strategy for combating cancer.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soon-Sang Kwon et al.
Molecules (Basel, Switzerland), 21(5) (2016-04-30)
Aschantin is a bioactive neolignan found in Magnolia flos with antiplasmodial, Ca(2+)-antagonistic, platelet activating factor-antagonistic, and chemopreventive activities. We investigated its inhibitory effects on the activities of eight major human cytochrome P450 (CYP) and uridine 5'-diphospho-glucuronosyltransferase (UGT) enzymes of human

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service