Skip to Content
Merck
All Photos(1)

Key Documents

SMB00907

Sigma-Aldrich

Nicotinamide riboside chloride

Nicotinamide riboside chloride

≥97% (HPLC), powder

Synonym(s):

Nicotinamide riboside chloride, Nicotinamide ribose chloride

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
R$917.00
250 MG
R$3,898.00

R$917.00


Estimated to ship onMarch 31, 2025


Request a Bulk Order

Select a Size

Change View
50 MG
R$917.00
250 MG
R$3,898.00

About This Item

Empirical Formula (Hill Notation):
C11H15N2O5Cl
CAS Number:
Molecular Weight:
290.70
UNSPSC Code:
12352205
NACRES:
NA.28

R$917.00


Estimated to ship onMarch 31, 2025


Request a Bulk Order

biological source

milk

Quality Level

Assay

≥97% (HPLC)

form

powder

color

white to off-white

solubility

10 mg/mL, clear, colorless

shipped in

wet ice

storage temp.

2-8°C

SMILES string

[Cl-].[n+]2(cc(ccc2)C(=O)N)[C@@H]1O[C@@H]([C@H]([C@H]1O)O)CO

InChI

1S/C11H14N2O5.ClH/c12-10(17)6-2-1-3-13(4-6)11-9(16)8(15)7(5-14)18-11;/h1-4,7-9,11,14-16H,5H2,(H-,12,17);1H/t7-,8-,9-,11-;/m1./s1

InChI key

YABIFCKURFRPPO-IVOJBTPCSA-N

General description

Nicotinamide riboside is involved in nicotinate and nicotinamide metabolism. Nicotinamide riboside was originally identified as a nutrient in milk and later discovered to be a NAD(+) precursor.

Application


  • Raising NAD(+) Level Stimulates Short-Chain Dehydrogenase/Reductase Proteins to Alleviate Heart Failure Independent of Mitochondrial Protein Deacetylation: This study demonstrates the potential of Nicotinamide Riboside Chloride to elevate NAD+ levels, thereby stimulating protein activity that can alleviate heart failure symptoms, highlighting its use as a cellular metabolism booster (Walker et al., 2023).

  • Acute Treatment with Nicotinamide Riboside Chloride Reduces Hippocampal Damage and Preserves the Cognitive Function of Mice with Ischemic Injury: This study investigates the neuroprotective effects of Nicotinamide Riboside Chloride, suggesting its potential in treating neurodegenerative diseases (Cheng et al., 2022).

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service