Skip to Content
Merck
All Photos(1)

Key Documents

SMB00802

Sigma-Aldrich

Bavachin

from Psoralea coryfolia, ≥97% (HPLC)

Synonym(s):

(2S)-2,3-Dihydro-7-hydroxy-2-(4-hydroxyphenyl)-6-(3-methyl-2-butenyl)-4H-1-benzopyran-4-one, 4′,7-Dihydroxy 6-(3-methyl 2-butenyl)flavanone, Corylifolin

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$1,289.00

R$1,289.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
R$1,289.00

About This Item

Empirical Formula (Hill Notation):
C20H20O4
CAS Number:
Molecular Weight:
324.37
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.28

R$1,289.00


Check Cart for Availability

Request a Bulk Order

Assay

≥97% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O1C(CC(=O)c3c1cc(c(c3)CC=C(C)C)O)c2ccc(cc2)O

InChI

1S/C20H20O4/c1-12(2)3-4-14-9-16-18(23)11-19(24-20(16)10-17(14)22)13-5-7-15(21)8-6-13/h3,5-10,19,21-22H,4,11H2,1-2H3

InChI key

OAUREGNZECGNQS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Bavachin is a flavonoid isolated from seeds of Psoralea corylifolia. It is a phytoestrogen that activates ERα and ERβ (EC50s = 320 and 680 nM, respectively), stimulates osteoblast proliferation and differentiation, and prevents bone loss following ovariectomy in rats.[1]

Bavachin is an inhibitor of acyl coenzyme A: cholesterol acyltransferase (ACAT). The IC50 value of bavachin was reported to be 86.0 μM in ACAT assay system using rat liver microsome.[2]

Bavachin is also reported to increase insulin-induced glucose uptake by differentiated adipocytes and myoblasts.[3]

Physical properties

Bavachin is soluble in DMSO at 30 mg/mL. Bavachin is not water soluble.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joonwoo Park et al.
Biomolecules & therapeutics, 20(2), 183-188 (2012-03-01)
In this study, we examined the estrogenic activity of bavachin, a component of Psoralea corylifolia that has been used as a traditional medicine in Asia. Bavachin was purified from ethanolic extract of Psoralea corylifolia and characterized its estrogenic activity by
Jung Ho Choi et al.
Archives of pharmacal research, 31(11), 1419-1423 (2008-11-22)
Acyl-coenzyme A: cholesterol acyltransferase (ACAT) catalyzes cholesterol esterification and plays important roles in intestinal absorption of cholesterol, hepatic production of lipoproteins and accumulation of cholesteryl ester within macrophages and smooth muscle cells. Ethanol extract of Psoralea corylifolia showed a significant
Hyejin Lee et al.
International journal of molecular sciences, 17(4), 527-527 (2016-04-14)
The fruit of Psoralea corylifolia L. (Fabaceae) (PC), known as "Bo-Gol-Zhee" in Korea has been used as traditional medicine. Ethanol and aqueous extracts of PC have an anti-hyperglycemic effect by increasing plasma insulin levels and decreasing blood glucose and total

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service