Skip to Content
Merck
All Photos(1)

Key Documents

SMB00243

Sigma-Aldrich

Sericic acid

≥95% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
R$2,548.00

R$2,548.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
R$2,548.00

About This Item

Empirical Formula (Hill Notation):
C30H48O6
CAS Number:
Molecular Weight:
504.70
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

R$2,548.00


Check Cart for Availability

Request a Bulk Order

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

CC1(C)CC[C@@]2(CC[C@]3(C)C(=CC[C@@H]4[C@@]5(C)C[C@@H](O)[C@H](O)[C@](C)(CO)[C@@H]5CC[C@@]34C)[C@@H]2[C@@H]1O)C(O)=O

InChI

1S/C30H48O6/c1-25(2)11-13-30(24(35)36)14-12-28(5)17(21(30)23(25)34)7-8-20-26(3)15-18(32)22(33)27(4,16-31)19(26)9-10-29(20,28)6/h7,18-23,31-34H,8-16H2,1-6H3,(H,35,36)/t18-,19-,20-,21-,22+,23+,26+,27-,28-,29-,30+/m1/s1

InChI key

IFIQVSCCFRXSJV-GOVAGAJPSA-N

General description

Natural product derived from plant source.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S C Hess et al.
Journal of ethnopharmacology, 47(2), 97-100 (1995-07-07)
Vochysia divergens Pohl (Vochysiaceae) is a tree commonly found in wet soils of 'Pantanal' of Mato Grosso, Brazil, and used in folk medicine against infections and asthma. We have studied different extracts and some isolated compounds from this plant for
Ouchen Wang et al.
PloS one, 6(6), e21130-e21130 (2011-06-23)
Natural products represent an important source for agents of cancer prevention and cancer treatment. More than 60% of conventional anticancer drugs are derived from natural sources, particularly from plant-derived materials. In this study, 2α, 3α, 19β, 23β-tetrahydroxyurs-12-en-28-oic acid (THA), a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service