Skip to Content
Merck
All Photos(1)

Key Documents

SCP0240

Sigma-Aldrich

Somatostatin 28 1-12

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
R$1,510.00
5 MG
R$4,713.00

R$1,510.00


Estimated to ship onMay 24, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
R$1,510.00
5 MG
R$4,713.00

About This Item

Empirical Formula (Hill Notation):
C49H81N17O19S1
Molecular Weight:
1244.33
UNSPSC Code:
51111800
NACRES:
NA.32

R$1,510.00


Estimated to ship onMay 24, 2025


Request a Bulk Order

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥85%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Ser-Ala-Asn-Ser-Asn-Pro-Ala-Met-Ala-Pro-Arg-Glu

Application

Somatostatin 28 1-12 (SS28(1-12)) is a somatostatin fragment that is monitored in brain tissue to track processing of somatostatin. SS28(1-12) may be used as an immunogen to generate SS28(1-12) antibodies.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service