All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C32H44N6O7
Molecular Weight:
624.73
UNSPSC Code:
12352200
NACRES:
NA.32
Recommended Products
Assay
≥95% (HPLC)
form
lyophilized
composition
Peptide Content, ≥85%
storage condition
protect from light
storage temp.
−20°C
Amino Acid Sequence
Asp-Pro-Ile-Leu-Trp
Application
JKC-301 (Asp-Pro-Ile-Leu-Trp) is a pentapeptide used as an selective endothelin ET(A) receptor antagonist.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
J E Tanus-Santos et al.
European journal of pharmacology, 396(1), 33-37 (2000-05-24)
The increased endothelin-1 levels observed after smoking may result from nicotine-stimulated endothelin-1 production by endothelial cells. In this study, we investigated the effects of selective endothelin ET(A) receptors antagonist Cycle D-a-aspartyl-L-prolyl-D-isoleucyl-D-tryptophyl (JKC 301) and of endothelin ET(B) receptors antagonist N-cis-2
Z Fekete et al.
The Journal of pharmacology and experimental therapeutics, 275(1), 215-218 (1995-10-01)
In this study we characterized the endothelin (ET) receptors of cultured L6 myotubes in order to gain a further insight into the mechanism of the ET effect on skeletal muscle cells. Displacements of 125I-ET-1 by unlabeled ET-1, ET-2 and ET-3
L C Ngoka et al.
Journal of mass spectrometry : JMS, 35(2), 265-276 (2000-02-19)
We previously showed by using mass spectrometry that endothelin A selective receptor antagonists BQ123 and JKC301 form novel coordination compounds with sodium ions. This property may underlie the ability of an ET(A) antagonist to induce net tubular sodium reabsorption in
Daphne Merkus et al.
American journal of physiology. Heart and circulatory physiology, 288(5), H2088-H2092 (2005-01-08)
Alpha-adrenergic vasoconstriction in the coronary circulation is mediated through alpha-adrenoceptors on cardiac myocytes and subsequent release of endothelin, a very potent, long-lasting vasoconstrictor. Recent studies found that adult cardiac myocytes do not express the preproendothelin gene. Thus we hypothesized that
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service