Skip to Content
Merck
All Photos(1)

Key Documents

SCP0129

Sigma-Aldrich

D-Val-Gly-Arg-pNA

≥95% (HPLC), lyophilized

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
R$1,569.00

R$1,569.00


Check Cart for Availability


Select a Size

Change View
10 MG
R$1,569.00

About This Item

Empirical Formula (Hill Notation):
C19H30N8O5
Molecular Weight:
450.49
UNSPSC Code:
12352202
NACRES:
NA.32

R$1,569.00


Check Cart for Availability

Product Name

D-Val-Gly-Arg-pNA,

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥70%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Val-Gly-Arg-pNA

Application

Val-Gly-Arg-pNA may be used as a chromogenic substrate of multicatalytic proteinase and peptidases and tissue plasminogen activator.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service