Skip to Content
Merck
All Photos(1)

Key Documents

SCP0080

Sigma-Aldrich

Caspase 2/3 Substrate

≥95% (HPLC), lyophilized

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$1,617.00

R$1,617.00


Estimated to ship onMay 26, 2025



Select a Size

Change View
5 MG
R$1,617.00

About This Item

Empirical Formula (Hill Notation):
C34H44N6O15
Molecular Weight:
776.74
UNSPSC Code:
12352202
NACRES:
NA.32

R$1,617.00


Estimated to ship onMay 26, 2025


Product Name

Caspase 2/3 Substrate,

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥80%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Ac-Leu-Asp-Glu-Ser-Asp-AMC

Application

Caspase 2/3 Substrate (Ac-LDESD-AMC) (Acetyl-Leu-Asp-Glu-Ser-Asp-amino-4-methylcoumarin) is a fluorogenic substrate that may be used to detect caspase 2 or caspase 3.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guoxing Fu et al.
Apoptosis : an international journal on programmed cell death, 13(11), 1291-1302 (2008-09-10)
Caspase-3, -6 and -7 cleave many proteins at specific sites to induce apoptosis. Their recognition of the P5 position in substrates has been investigated by kinetics, modeling and crystallography. Caspase-3 and -6 recognize P5 in pentapeptides as shown by enzyme
Andreas Schweizer et al.
The Journal of biological chemistry, 278(43), 42441-42447 (2003-08-16)
The cell death protease caspase-2 has recently been recognized as the most apical caspase in the apoptotic cascade ignited during cell stress signaling. Cytotoxic stress, such as that caused by cancer therapies, leads to activation of caspase-2, which acts as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service