Skip to Content
Merck
All Photos(1)

Key Documents

SCP0034

Sigma-Aldrich

Amyloid β 12-28 human

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
R$1,030.00
1 MG
R$1,710.00

R$1,030.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
500 μG
R$1,030.00
1 MG
R$1,710.00

About This Item

Empirical Formula (Hill Notation):
C89H135N25O25
Molecular Weight:
1955.18
UNSPSC Code:
12352200
NACRES:
NA.32

R$1,030.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥70%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys

Application

Amyloid β (Aβ) refers to peptides derived from Amyloid precursor protein that vary in length from 36-43 amino acids. Aβ(s) peptides, their peptide fragments and mutated fragments are used to study a wide range of metabolic and regulatory functions including activation of kinases, regulation of cholesterol transport, function as a transcription factor, and regulators of inflammation. Aβ(s) peptides and their peptide fragments are also used to study oxidative stress, metal binding and mechanisms of protein cross-linking in the context of diseases such as Alzheimer′s disease and neurodegeneration.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samaneh Beheshti et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 13(2), 542-548 (2011-12-14)
The synthesis and characterization of four new Ferrocene (Fc) bioconjugates, bearing a podant (Lys)-Leu-Val-Phe-Phe motif, namely the hydrophobic sequence of amyloid-β-peptides (Aβ), is reported. The Fc-peptide conjugates are characterized by a reversible redox activity and the ability to undergo hydrophobic
Zanxia Cao et al.
International journal of molecular sciences, 12(11), 8259-8274 (2011-12-17)
The aim of this work is to investigate the effects of different force fields and temperatures on the structural character of Aβ (12-28) peptide in aqueous solution. Moreover, the structural character of Aβ (12-28) peptide is compared with other amyloid
Marino Convertino et al.
The Journal of biological chemistry, 286(48), 41578-41588 (2011-10-05)
In recent years, an increasing number of small molecules and short peptides have been identified that interfere with aggregation and/or oligomerization of the Alzheimer β-amyloid peptide (Aβ). Many of them possess aromatic moieties, suggesting a dominant role for those in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service