Skip to Content
Merck
All Photos(1)

Documents

SBR00088

Sigma-Aldrich

Rilpivirine

new

≥95% (HPLC), powder

Synonym(s):

4-[[4-[[4-[(1E)-2-Cyanoethenyl]-2,6-dimethylphenyl]amino]-2-pyrimidinyl]amino]benzonitrile, R 278474, TMC 278

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H18N6
CAS Number:
Molecular Weight:
366.42
MDL number:
UNSPSC Code:
51101500

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to yellow, or beige

antibiotic activity spectrum

viruses

Mode of action

enzyme | inhibits

shipped in

ambient

storage temp.

−20°C

SMILES string

CC1=CC(=CC(=C1NC2=NC(=NC=C2)NC3=CC=C(C=C3)C#N)C)/C=C/C#N

InChI

1S/C22H18N6/c1-15-12-18(4-3-10-23)13-16(2)21(15)27-20-9-11-25-22(28-20)26-19-7-5-17(14-24)6-8-19/h3-9,11-13H,1-2H3,(H2,25,26,27,28)/b4-3+

InChI key

YIBOMRUWOWDFLG-ONEGZZNKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Rilpivirine is an antibiotic which belongs to the class of compounds known as non-nucleoside reverse transcriptase inhibitors (NNRTIs).

Biochem/physiol Actions

Rilpivirine inhibits the activity of the reverse transcriptase enzyme, which is essential for the replication of HIV-1. Specifically, it binds to a specific site on the enzyme and blocks its activity, thereby preventing the conversion of viral RNA into DNA, which is an essential step for the integration of viral DNA into host cells.
Rilpivirine is a next generation nonnucleoside reverse transcriptase inhibitor (NNRTI), It has been shown to be active against many HIV strains with mutations that confer resistance to the first-generation NNRTIs.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service