Skip to Content
Merck
All Photos(1)

Key Documents

SBR00037

Sigma-Aldrich

Solithromycin

≥98% (HPLC)

Synonym(s):

2H-Oxacyclotetradecino[4,3-d]oxazole-2,6,8,14(1H,7H,9H)-tetrone, CEM-101, OP-1068

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
R$500.00

R$500.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
R$500.00

About This Item

Empirical Formula (Hill Notation):
C43H65FN6O10
Molecular Weight:
845.01
MDL number:
UNSPSC Code:
51102829
NACRES:
NA.76

R$500.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

protein synthesis | interferes

storage temp.

−20°C

SMILES string

F[C@@]1(C(=O)O[C@@H]([C@]2(OC(=O)N([C@@H]2[C@H](C(=O)[C@@H](C[C@]([C@@H]([C@H](C1=O)C)O[C@@H]5O[C@@H](C[C@@H]([C@H]5O)N(C)C)C)(OC)C)C)C)CCCC[n]3nnc(c3)c4cc(ccc4)N)C)CC)C

InChI

1S/C43H65FN6O10/c1-12-32-43(8)35(50(40(55)60-43)19-14-13-18-49-23-30(46-47-49)28-16-15-17-29(45)21-28)26(4)33(51)24(2)22-41(6,56-11)37(27(5)36(53)42(7,44)39(54)58-32)59-38-34(52)31(48(9)10)20-25(3)57-38/h15-17,21,23-27,31-32,34-35,37-38,52H,12-14,18-20,22,45H2,1-11H3/t24-,25-,26+,27+,31+,32-,34-,35-,37-,38+,41-,42+,43-/m1/s1

InChI key

IXXFZUPTQVDPPK-ZAWHAJPISA-N

General description

Solithromycin is a highly potent next-generation macrolid. the first fluoroketolid. which has potent activity against most macrolide-resistant strains. Solithromycin (CEM-101) is a novel fluoroketolide with improved antimicrobial effectiveness.

Application

Solithromycin has been shown to have potentent activity against S. pneumoniae as well as an extended spectrum of activity against CA-MRS. enterococc according to in vitro and in vivo research. Solithromycin has been used in:

  • the research on the anti-inflammatory effects of macrolides[1]
  • the research of potency of solithromycin in comparison to other antimicrobials[2]
  • in ithe research of the vitro activity of solithromycin against clinical N. gonorrhoeae isolates and reference strain[3]

Biochem/physiol Actions

Mode of Action: Solithromycin binds to the large 50S subunit of the ribosome and inhibits protein biosynthesis. Like other ketolide. it should impair bacterial ribosomal subunit formation.

Antimicrobial spectrum: Active against bacteria like Legionella, Chlamydophila, Chlamydia, Mycoplasma, and Ureaplasma and against gonococci and other organisms that cause genitourinary tract infections

Features and Benefits

High quality antibiotic suitable for mulitple research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In vitro activity of the new fluoroketolide solithromycin (CEM-101) against a large collection of clinical Neisseria gonorrhoeae isolates and international reference strains, including those with high-level antimicrobial resistance: potential treatment option for gonorrhea?
Golparian et al.
Antimicrobial Agents and Chemotherapy, 56, 2739-2742 (2012)
Ward Rodgers et al.
Antimicrobial agents and chemotherapy, 57(4), 1632-1637 (2013-01-16)
The continuing increase in antibiotic-resistant microorganisms is driving the search for new antibiotic targets and improved antimicrobial agents. Ketolides are semisynthetic derivatives of macrolide antibiotics, which are effective against certain resistant organisms. Solithromycin (CEM-101) is a novel fluoroketolide with improved
Antimicrobial characterisation of solithromycin (CEM-101), a novel fluoroketolide: activity against staphylococci and enterococci
International Journal of Antimicrobial Agents, 37, 39-45 (2010)
Yoshiki Kobayashi et al.
The Journal of pharmacology and experimental therapeutics, 345(1), 76-84 (2013-01-30)
Macrolides are reported to reduce exacerbation of chronic inflammatory respiratory disease, such as chronic obstructive pulmonary disease (COPD), and also show anti-inflammatory effects in vitro and in vivo. However the anti-inflammatory efficacies of current macrolides are relatively weak. Here we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service