Select a Size
R$381.00
Estimated to ship onMay 27, 2025
Select a Size
About This Item
R$381.00
Estimated to ship onMay 27, 2025
biological source
synthetic
form
liquid
species reactivity
rat, mouse, human
concentration
200 μg/mL
shipped in
wet ice
storage temp.
−20°C
Immunogen
Application
Linkage
Disclaimer
related product
Storage Class Code
10 - Combustible liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service