Skip to Content
Merck
All Photos(1)

Key Documents

SBP3500736

Sigma-Aldrich

DBX1 Blocking Peptide for SAB3500736

Sign Into View Organizational & Contract Pricing

Select a Size

0.05 MG
R$381.00

R$381.00


Estimated to ship onJune 02, 2025



Select a Size

Change View
0.05 MG
R$381.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$381.00


Estimated to ship onJune 02, 2025


biological source

synthetic

form

liquid

species reactivity

human, mouse, rat

concentration

200 μg/mL

shipped in

wet ice

storage temp.

−20°C

Immunogen

15 amino acids near the carboxy terminus of human DBX1.

Application

Used as a blocking peptide in immunoblotting applications.

Linkage

This peptide blocks the action of antibody SAB3500736.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

related product

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service