Skip to Content
Merck
All Photos(1)

Key Documents

SBP3500580

Sigma-Aldrich

APBA2 Blocking Peptide for SAB3500580

Sign Into View Organizational & Contract Pricing

Select a Size

0.05 MG
R$381.00

R$381.00


Check Cart for Availability


Select a Size

Change View
0.05 MG
R$381.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$381.00


Check Cart for Availability

biological source

synthetic

form

liquid

species reactivity

human

concentration

200 μg/mL

shipped in

wet ice

storage temp.

−20°C

Immunogen

18 amino acids near the amino terminus of human APBA2.

Application

Used as a blocking peptide in immunoblotting applications.

Linkage

This peptide blocks the action of antibody SAB3500580.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service