Skip to Content
Merck
All Photos(1)

Key Documents

SAE0063

Sigma-Aldrich

Alkaline Phosphatase Recombinant

solution (high-activity)

Synonym(s):

ALP

Sign Into View Organizational & Contract Pricing

Select a Size

15 MG
R$8,250.00

R$8,250.00


Check Cart for Availability


Select a Size

Change View
15 MG
R$8,250.00

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54

R$8,250.00


Check Cart for Availability

recombinant

expressed in proprietary host

Quality Level

form

solution (high-activity)

shipped in

wet ice

storage temp.

2-8°C

General description

This high-activity recombinant alkaline phosphatase has a minimum Specific Activity of ≥ 15,000 U/mg (measured at 37 °C), and a minimum purity of ≥ 95% by HPLC.

No animal-derived components are used in the production process. This mitigates the risk of Bovine Spongiform Encephalopathy (BSE) and other animal-caused infections. This animal component free (ACF) recombinant alkaline phosphatase guarantees superior lot-to-lot consistency and eliminates BSE risk.

Application

Alkaline phosphatase is used for conjugation to antibodies and other proteins for ELISA, Western blotting, and histochemical detection.

Unit Definition

One unit hydrolyzes 1.0 μmole of p-nitrophenyl phosphate per minute at pH 9.8 (DEA) at 37 deg C.

Physical form

Solution in 3 M NaCl with triethanolamine, pH 7.5

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service